siRNA Details
| Gene Symbol: Ocstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029021.1 | NP_083297.1 | 3 | 1420 | ||
| GenBank mRNA | AK014751.1 | - | 3 | 1420 | ||
| AK042271.1 | - | 3 | 1432 | |||
| BC020160.1 | - | 3 | 1388 | 3966086 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ocstamp| Gene Symbol: Ocstamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029021.1 | NP_083297.1 | 3 | 1420 | ||
| GenBank mRNA | AK014751.1 | - | 3 | 1420 | ||
| AK042271.1 | - | 3 | 1432 | |||
| BC020160.1 | - | 3 | 1388 | 3966086 | ||