siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127191.2 | NP_001120663.1 | 4, 5 | 920 | ||
| NM_029068.4 | NP_083344.3 | 4, 5 | 920 | |||
| GenBank mRNA | AK015872.1 | - | 4, 5 | 881 | ||
| AK032055.1 | - | 4, 5 | 877 | |||
| AK166422.1 | - | 4, 5 | 894 | |||
| AK215008.1 | - | 1, 2 | 73 | |||
| BC019424.1 | - | 3, 4 | 685 | 5052234 | ||
| BC070431.1 | - | 863 | ||||

Snx16
Genomic Map