siRNA Details
| Gene Symbol: 1700012B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029306.3 | NP_083582.2 | 4 | 541 | ||
| GenBank mRNA | AK005747.1 | - | 4 | 542 | ||
| BC092535.1 | - | 4 | 503 | 6773440 | ||
| CF105954.1 | - | 4 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012B09Rik| Gene Symbol: 1700012B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029306.3 | NP_083582.2 | 4 | 541 | ||
| GenBank mRNA | AK005747.1 | - | 4 | 542 | ||
| BC092535.1 | - | 4 | 503 | 6773440 | ||
| CF105954.1 | - | 4 | 490 | |||