siRNA Details
Gene Symbol: Spata9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029343.3 | NP_083619.1 | 5 | 817 | ||
GenBank mRNA | AK006554.1 | - | 5 | 818 | ||
BC048558.1 | - | 5 | 797 | 6703288 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spata9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029343.3 | NP_083619.1 | 5 | 817 | ||
GenBank mRNA | AK006554.1 | - | 5 | 818 | ||
BC048558.1 | - | 5 | 797 | 6703288 |