siRNA Details
| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029394.3 | NP_083670.1 | 3 | 368 | ||
| GenBank mRNA | AK012711.1 | - | 3 | 260 | ||
| AK017610.1 | - | 3 | 262 | |||
| BC051168.1 | - | 3 | 371 | 6306904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx24| Gene Symbol: Snx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029394.3 | NP_083670.1 | 3 | 368 | ||
| GenBank mRNA | AK012711.1 | - | 3 | 260 | ||
| AK017610.1 | - | 3 | 262 | |||
| BC051168.1 | - | 3 | 371 | 6306904 | ||