siRNA Details
| Gene Symbol: Atp11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029570.3 | NP_083846.2 | 28, 29 | 3563 | ||
| GenBank mRNA | AK003820.1 | - | 1, 2 | 464 | ||
| AK129248.1 | - | 28, 29 | 3555 | |||
| BC026741.1 | - | 2, 3 | 284 | |||
| BC026986.1 | - | 13, 14 | 1632 | |||
| BC076603.1 | - | 28, 29 | 3433 | 30629594 | ||
| CF952078.1 | - | 1, 2 | 177 | |||

Atp11b
Genomic Map