siRNA Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029624.4 | NP_083900.3 | 6, 7 | 918 | ||
| GenBank mRNA | AK010312.1 | - | 6, 7 | 889 | ||
| AK041659.1 | - | 4, 5 | 585 | |||
| AK154622.1 | - | 6, 7 | 915 | |||
| AK169983.1 | - | 6, 7 | 916 | |||
| AK177501.1 | - | 1, 2 | 50 | |||
| AK183502.1 | - | 1, 2 | 50 | |||
| BC020104.1 | - | 6, 7 | 925 | 3963699 | ||

Lmf1
Genomic Map