siRNA Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290679.1 | NP_001277608.1 | 4, 5 | 599 | ||
| GenBank mRNA | AK017734.1 | - | 4, 5 | 514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem14a| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290679.1 | NP_001277608.1 | 4, 5 | 599 | ||
| GenBank mRNA | AK017734.1 | - | 4, 5 | 514 | ||