siRNA Details
| Gene Symbol: Wdr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029453.2 | NP_083729.2 | 15 | 1978 | ||
| GenBank mRNA | AK015753.1 | - | 4 | 431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr64| Gene Symbol: Wdr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029453.2 | NP_083729.2 | 15 | 1978 | ||
| GenBank mRNA | AK015753.1 | - | 4 | 431 | ||