siRNA Details
Gene Symbol: Vmn2r-ps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007132.1 | - | 4 | 443 | ||
AK016194.1 | - | 3 | 536 | |||
AK030272.1 | - | 3 | 503 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn2r-ps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007132.1 | - | 4 | 443 | ||
AK016194.1 | - | 3 | 536 | |||
AK030272.1 | - | 3 | 503 |