siRNA Details
| Gene Symbol: NOTCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000435.2 | NP_000426.2 | 5 | 771 | ||
| XM_005259924.4 | XP_005259981.1 | 5 | 710 | |||
| GenBank mRNA | AB209447.1 | - | 5 | 704 | ||
| U97669.1 | - | 5 | 773 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOTCH3| Gene Symbol: NOTCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000435.2 | NP_000426.2 | 5 | 771 | ||
| XM_005259924.4 | XP_005259981.1 | 5 | 710 | |||
| GenBank mRNA | AB209447.1 | - | 5 | 704 | ||
| U97669.1 | - | 5 | 773 | |||