siRNA Details
| Gene Symbol: Zfp60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018216.1 | - | 1151 | |||
| BC094367.1 | - | 801 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp60| Gene Symbol: Zfp60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018216.1 | - | 1151 | |||
| BC094367.1 | - | 801 | ||||