siRNA Details
| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029585.3 | NP_083861.1 | 2 | 690 | ||
| GenBank mRNA | AK011666.1 | - | 2 | 716 | ||
| BC031930.1 | - | 2 | 690 | 4018106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Det1| Gene Symbol: Det1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029585.3 | NP_083861.1 | 2 | 690 | ||
| GenBank mRNA | AK011666.1 | - | 2 | 716 | ||
| BC031930.1 | - | 2 | 690 | 4018106 | ||