siRNA Details
| Gene Symbol: 1700012A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029587.2 | NP_083863.1 | 3 | 518 | ||
| GenBank mRNA | AK005884.1 | - | 3 | 518 | ||
| BC059718.1 | - | 3 | 217 | |||
| BC087908.1 | - | 3 | 501 | 6774051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700012A03Rik| Gene Symbol: 1700012A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029587.2 | NP_083863.1 | 3 | 518 | ||
| GenBank mRNA | AK005884.1 | - | 3 | 518 | ||
| BC059718.1 | - | 3 | 217 | |||
| BC087908.1 | - | 3 | 501 | 6774051 | ||