siRNA Details
| Gene Symbol: Ift43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199843.1 | NP_001186772.1 | 6 | 358 | ||
| NM_029601.3 | NP_083877.1 | 5 | 325 | |||
| GenBank mRNA | AK006115.1 | - | 5 | 323 | ||
| BC034668.1 | - | 6 | 296 | |||
| BC038915.1 | - | 6 | 347 | 5148723 | ||
| CF751010.1 | - | 6 | 321 | |||

Ift43
Genomic Map