siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029391.2 | NP_083667.1 | 6, 7 | 675 | ||
| GenBank mRNA | AB232592.1 | - | 6, 7 | 534 | ||
| AF408432.1 | - | 6, 7 | 647 | |||
| AK005314.1 | - | 6, 7 | 675 | |||
| AK140513.1 | - | 5, 6 | 872 | |||
| AK170552.1 | - | 6, 7 | 689 | |||
| AK180450.1 | - | 1, 2 | 185 | |||
| AK219531.1 | - | 2, 3 | 224 | |||
| BC007147.1 | - | 6, 7 | 619 | 3600193 | ||
| BC096654.1 | - | 6, 7 | 658 | 5056766 | ||

Rab4b
Genomic Map