siRNA Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290679.1 | NP_001277608.1 | 5 | 1286 | ||
| NM_029398.3 | NP_083674.2 | 5 | 1103 | |||
| GenBank mRNA | AK017734.1 | - | 5 | 1200 | ||
| BC060034.1 | - | 5 | 1047 | 6491675 | ||

Tmem14a
Genomic Map