siRNA Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029413.4 | NP_083689.2 | 16 | 3148 | ||
| GenBank mRNA | AK032807.1 | - | 16 | 3149 | ||
| BC132153.1 | - | 16 | 3002 | 40130430 | ||
| BC132497.1 | - | 16 | 3002 | 40130774 | ||

Morc4
Genomic Map