siRNA Details
| Gene Symbol: Klf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029416.2 | NP_083692.2 | 3 | 2192 | ||
| GenBank mRNA | AF508984.1 | - | 3 | 2159 | ||
| AK018361.1 | - | 3 | 2158 | |||
| AK139827.1 | - | 3 | 2159 | |||
| BC064748.1 | - | 3 | 2136 | 30476790 | ||

Klf17
Genomic Map