siRNA Details
| Gene Symbol: Lca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027448.2 | NP_081724.1 | 4 | 1176 | ||
| NM_029434.3 | NP_083710.2 | 4 | 1072 | |||
| GenBank mRNA | AK015260.1 | - | 4 | 1044 | ||
| BC049101.1 | - | 4 | 1104 | 5702365 | ||
| BC052060.1 | - | 4 | 1104 | 5702365 | ||

Lca5
Genomic Map