siRNA Details
| Gene Symbol: Tcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029467.3 | NP_083743.2 | 5 | 1134 | ||
| GenBank mRNA | AK016263.1 | - | 5 | 1004 | ||
| AK076581.1 | - | 5 | 1055 | |||
| BC050746.1 | - | 1058 | 6741794 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcam1| Gene Symbol: Tcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029467.3 | NP_083743.2 | 5 | 1134 | ||
| GenBank mRNA | AK016263.1 | - | 5 | 1004 | ||
| AK076581.1 | - | 5 | 1055 | |||
| BC050746.1 | - | 1058 | 6741794 | |||