siRNA Details
| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029472.3 | NP_083748.3 | 2 | 302 | ||
| GenBank mRNA | AK016340.1 | - | 2 | 286 | ||
| BC052144.1 | - | 2 | 311 | |||
| BC061011.1 | - | 2 | 302 | 6774382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstt4| Gene Symbol: Gstt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029472.3 | NP_083748.3 | 2 | 302 | ||
| GenBank mRNA | AK016340.1 | - | 2 | 286 | ||
| BC052144.1 | - | 2 | 311 | |||
| BC061011.1 | - | 2 | 302 | 6774382 | ||