siRNA Details
| Gene Symbol: Zdhhc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029492.4 | NP_083768.4 | 4, 5 | 459 | ||
| GenBank mRNA | AK017158.1 | - | 4, 5 | 460 | ||
| AK031184.1 | - | 4, 5 | 418 | |||
| AK049270.1 | - | 4, 5 | 439 | |||
| AK140182.1 | - | 4, 5 | 420 | |||
| AK151840.1 | - | 4, 5 | 474 | |||
| AK163174.1 | - | 4, 5 | 457 | |||
| AK170017.1 | - | 4, 5 | 477 | |||
| AK170139.1 | - | 4, 5 | 400 | |||
| AY668951.1 | - | 4, 5 | 377 | |||
| BC019536.1 | - | 4, 5 | 476 | 4039734 | ||

Zdhhc20
Genomic Map