siRNA Details
| Gene Symbol: Stambpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029682.4 | NP_083958.3 | 8 | 1425 | ||
| GenBank mRNA | AB010121.1 | - | 8 | 1120 | ||
| AB010122.1 | - | 8 | 1084 | |||
| AB066211.1 | - | 8 | 1425 | |||
| AB066212.1 | - | 4 | 774 | |||
| AB066213.1 | - | 8 | 1108 | |||
| AK007087.1 | - | 4 | 758 | |||
| BC018343.1 | - | 8 | 1151 | 4010423 | ||
| BC023097.1 | - | 8 | 1394 | |||

Stambpl1
Genomic Map