siRNA Details
| Gene Symbol: Catsper3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252487.1 | NP_001239416.1 | 1 | 169 | ||
| NM_001252488.1 | NP_001239417.1 | 1 | 169 | |||
| NM_029772.4 | NP_084048.1 | 1 | 169 | |||
| GenBank mRNA | AK014942.1 | - | 1 | 170 | ||
| AY263400.1 | - | 1 | 144 | |||
| HM179540.1 | - | 1 | 86 | |||
| HM179541.1 | - | 1 | 86 | |||

Catsper3
Genomic Map