siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001326.1 | NP_001001326.1 | 6 | 1796 | ||
| NM_029811.2 | NP_084087.2 | 5 | 545 | |||
| GenBank mRNA | AK158270.1 | - | 6 | 1801 | ||
| BC030391.1 | - | 2 | 271 | 4950637 | ||
| BC051034.1 | - | 6 | 1977 | 5345299 | ||
| CF737707.1 | - | 3 | 278 | |||

St5
Genomic Map