siRNA Details
| Gene Symbol: 2510039O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029841.3 | NP_084117.2 | 1 | 852 | ||
| GenBank mRNA | AK011059.1 | - | 1 | 852 | ||
| AK153761.1 | - | 1 | 853 | |||
| AK173332.1 | - | 2 | 707 | |||
| BC012878.1 | - | 1 | 825 | 4190292 | ||

2510039O18Rik
Genomic Map