siRNA Details
| Gene Symbol: Ermn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029972.3 | NP_084248.2 | 1 | 137 | ||
| GenBank mRNA | AK020733.1 | - | 1 | 249 | ||
| AK078089.1 | - | 1 | 227 | |||
| AK136820.1 | - | 1 | 227 | |||
| AK137305.1 | - | 1 | 229 | |||
| AK220372.1 | - | 1 | 256 | |||
| BC089515.1 | - | 1 | 138 | 6589784 | ||
| BC090670.1 | - | 1 | 169 | 30941185 | ||
| DQ334270.1 | - | 1 | 115 | |||

Ermn
Genomic Map