siRNA Details
| Gene Symbol: Plet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029639.2 | NP_083915.2 | 2, 3 | 436 | ||
| GenBank mRNA | AK002767.1 | - | 2, 3 | 438 | ||
| AK005558.1 | - | 2, 3 | 437 | |||
| AY364436.1 | - | 2, 3 | 436 | |||
| BC017624.1 | - | 2, 3 | 420 | 4487871 | ||
| BC022950.1 | - | 2, 3 | 416 | 5148408 | ||

Plet1
Genomic Map