siRNA Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252437.1 | NP_001239366.1 | 2 | 448 | ||
| NM_029657.4 | NP_083933.1 | 2 | 445 | |||
| GenBank mRNA | AK011747.2 | - | 2 | 238 | ||
| AK088533.1 | - | 2 | 228 | |||
| AK150176.1 | - | 3 | 281 | |||
| AK153407.1 | - | 2 | 269 | |||
| AK192041.1 | - | 1 | 73 | |||
| AK211892.1 | - | 1 | 38 | |||
| BC046830.1 | - | 2 | 294 | 6811606 | ||

Mgrn1
Genomic Map