siRNA Details
Gene Symbol: Pkd1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029686.4 | NP_083962.4 | 1 | 232 | ||
GenBank mRNA | AY164484.1 | - | 1 | 232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pkd1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029686.4 | NP_083962.4 | 1 | 232 | ||
GenBank mRNA | AY164484.1 | - | 1 | 232 |