siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 2 | 319 | ||
| GenBank mRNA | AK012236.1 | - | 2 | 253 | ||
| AK089920.1 | - | 2 | 303 | |||
| AK157348.1 | - | 2 | 281 | |||
| AK169298.1 | - | 2 | 289 | |||
| AK171726.1 | - | 2 | 238 | |||
| BC011435.1 | - | 2 | 259 | 4010102 | ||
| CJ093818.1 | - | 2 | 319 | |||

Cog2
Genomic Map