siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 9, 10 | 1417 | ||
| GenBank mRNA | AK012392.1 | - | 9, 10 | 1205 | ||
| AK051683.1 | - | 9, 10 | 1220 | |||
| AK132316.1 | - | 9, 10 | 1213 | |||
| AK133622.1 | - | 9, 10 | 1161 | |||
| AK136921.1 | - | 9, 10 | 1240 | |||

Sdccag8
Genomic Map