siRNA Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146119.1 | NP_001139591.1 | 5 | 532 | ||
| NM_029758.4 | NP_084034.2 | 6 | 605 | |||
| GenBank mRNA | AK010821.1 | - | 4 | 468 | ||
| AK047142.1 | - | 5 | 495 | |||
| BC049664.1 | - | 6 | 567 | 6532813 | ||
| BC052465.1 | - | 5 | 513 | 6834768 | ||

Fam49a
Genomic Map