siRNA Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000349.2 | NP_000340.2 | 4 | 667 | ||
| XM_006716392.2 | XP_006716455.1 | 4 | 850 | |||
| GenBank mRNA | AF035277.1 | - | 4 | 515 | ||
| AK090513.1 | - | 3 | 851 | |||
| AK315122.1 | - | 4 | 537 | |||
| AL526126.3 | - | 4 | 674 | |||
| AV708514.1 | - | 2 | 155 | |||
| BC010550.1 | - | 4 | 468 | 4153601 | ||
| CR456803.1 | - | 4 | 403 | |||
| CR541850.1 | - | 4 | 403 | |||
| KU178337.1 | - | 4 | 403 | |||
| KU178338.1 | - | 4 | 403 | |||
| S79669.1 | - | 4 | 555 | |||
| U17280.1 | - | 4 | 529 | |||

STAR
Genomic Map