siRNA Details
| Gene Symbol: 4632433K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014606.1 | - | 836 | |||
| BC147185.1 | - | 796 | 9056235 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4632433K11Rik| Gene Symbol: 4632433K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014606.1 | - | 836 | |||
| BC147185.1 | - | 796 | 9056235 | |||