siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029852.2 | NP_084128.2 | 2 | 510 | ||
| GenBank mRNA | AK011135.1 | - | 2 | 582 | ||
| AK015007.1 | - | 2 | 647 | |||
| AK141627.1 | - | 2 | 631 | |||
| BC028960.1 | - | 1 | 211 | |||
| BC043468.1 | - | 1 | 124 | |||
| BC070464.1 | - | 2 | 510 | 30608497 | ||

Cep83
Genomic Map