siRNA Details
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 11 | 1633 | ||
GenBank mRNA | AK008613.1 | - | 9 | 1286 | ||
AK008663.1 | - | 11 | 1629 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 11 | 1633 | ||
GenBank mRNA | AK008613.1 | - | 9 | 1286 | ||
AK008663.1 | - | 11 | 1629 |