siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 12 | 1768 | ||
| GenBank mRNA | AK051683.1 | - | 13 | 1779 | ||
| AK132316.1 | - | 12 | 1564 | |||
| AK133622.1 | - | 12 | 1512 | |||
| AK136921.1 | - | 13 | 1799 | |||
| BC016244.1 | - | 13 | 1663 | 4507575 | ||

Sdccag8
Genomic Map