siRNA Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290649.1 | NP_001277578.1 | 7 | 808 | ||
| NM_029793.2 | NP_084069.1 | 8 | 883 | |||
| GenBank mRNA | AK002954.1 | - | 8 | 827 | ||
| AK008972.1 | - | 8 | 782 | |||
| AK028516.1 | - | 8 | 802 | |||
| AK044987.1 | - | 8 | 831 | |||
| BC024637.1 | - | 8 | 805 | |||
| BC037037.1 | - | 4 | 320 | |||
| BC043090.1 | - | 8 | 748 | 6418353 | ||
| BC053068.1 | - | 7 | 736 | 6405595 | ||

Golga1
Genomic Map