siRNA Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109913.1 | NP_001103383.1 | 2 | 230 | ||
| NM_029804.3 | NP_084080.1 | 2 | 230 | |||
| GenBank mRNA | AK011521.1 | - | 2 | 176 | ||
| AK051976.1 | - | 2 | 176 | |||
| AK133911.1 | - | 2 | 165 | |||
| AK145343.1 | - | 2 | 168 | |||
| AK159848.1 | - | 2 | 162 | |||
| AK166670.1 | - | 2 | 165 | |||
| AK168347.1 | - | 2 | 165 | |||
| AK169101.1 | - | 2 | 165 | |||
| AK171892.1 | - | 2 | 165 | |||
| AK209547.1 | - | 1 | 38 | |||
| AK220299.1 | - | 2 | 190 | |||
| BC017539.1 | - | 2 | 160 | |||
| BC035525.1 | - | 2 | 165 | |||
| BC054785.1 | - | 1632 | ||||
| BC065172.1 | - | 2 | 160 | 6415170 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109911, NM_053876, XM_006241116, U32577, AY724543, and others.

Hnrnpm
Genomic Map