siRNA Details
| Gene Symbol: 2510039O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029841.3 | NP_084117.2 | 3 | 2317 | ||
| GenBank mRNA | AK011059.1 | - | 3 | 2317 | ||
| AK173332.1 | - | 3 | 3311 | |||
| AK183513.1 | - | 1 | 217 | |||
| AK195992.1 | - | 1 | 216 | |||
| AK197588.1 | - | 1 | 216 | |||
| AK202703.1 | - | 1 | 216 | |||
| AK209381.1 | - | 1 | 217 | |||
| AK212923.1 | - | 1 | 200 | |||
| BC012878.1 | - | 3 | 2290 | 4190292 | ||

2510039O18Rik
Genomic Map