siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 2 | 242 | ||
| GenBank mRNA | AK021042.1 | - | 2 | 133 | ||
| AY102701.1 | - | 2 | 164 | |||
| BC027201.1 | - | 2 | 178 | |||
| BC043701.1 | - | 2 | 155 | |||
| BC056435.1 | - | 2 | 151 | 5707801 | ||
| BY100294.1 | - | 2 | 257 | |||

Naa35
Genomic Map