siRNA Details
| Gene Symbol: Spata2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030176.2 | NP_084452.2 | 2 | 382 | ||
| GenBank mRNA | AK019166.1 | - | 1 | 707 | ||
| AK081234.1 | - | 2 | 382 | |||
| AK139339.1 | - | 681 | ||||
| AK170267.1 | - | 1 | 682 | |||
| AK220503.1 | - | 682 | ||||
| BC139310.1 | - | 1 | 317 | 8862332 | ||
| BC139311.1 | - | 1 | 317 | 8862333 | ||

Spata2l
Genomic Map