siRNA Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271483.1 | NP_001258412.1 | 2 | 234 | ||
| NM_001271484.1 | NP_001258413.1 | 2 | 319 | |||
| NM_030179.3 | NP_084455.2 | 2 | 222 | |||
| NM_175378.1 | NP_780587.1 | 2 | 319 | |||
| GenBank mRNA | AK018895.1 | - | 2 | 312 | ||
| AK019516.1 | - | 2 | 235 | |||
| AK029962.1 | - | 2 | 320 | |||
| AK161301.1 | - | 2 | 309 | |||
| BC035226.1 | - | 2 | 222 | 4920788 | ||

Clip4
Genomic Map