siRNA Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030189.3 | NP_084465.1 | 2, 3 | 300 | ||
| GenBank mRNA | AK015061.1 | - | 3, 4 | 495 | ||
| AK019766.1 | - | 2, 3 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stpg1| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030189.3 | NP_084465.1 | 2, 3 | 300 | ||
| GenBank mRNA | AK015061.1 | - | 3, 4 | 495 | ||
| AK019766.1 | - | 2, 3 | 301 | |||