siRNA Details
| Gene Symbol: Gas2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190406.1 | NP_001177335.1 | 2 | 687 | ||
| NM_001190408.1 | NP_001177337.1 | 2 | 687 | |||
| NM_030228.3 | NP_084504.2 | 3 | 945 | |||
| NM_144560.3 | NP_653146.1 | 3 | 945 | |||
| GenBank mRNA | AF508323.1 | - | 1 | 589 | ||
| AF508324.1 | - | 1 | 589 | |||
| AK019661.1 | - | 3 | 920 | |||
| AK052229.1 | - | 3 | 946 | |||
| AK082864.1 | - | 2 | 655 | |||
| AK218035.1 | - | 1 | 60 | |||
| BC031785.1 | - | 2 | 637 | 4194333 | ||

Gas2l1
Genomic Map