siRNA Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146085.1 | NP_001139557.1 | 3, 4 | 352 | ||
| NM_001146086.1 | NP_001139558.1 | 2, 3 | 275 | |||
| NM_030236.3 | NP_084512.2 | 1, 2 | 165 | |||
| GenBank mRNA | AK013711.1 | - | 5, 6 | 714 | ||
| AK020018.1 | - | 1, 2 | 166 | |||
| AK171630.1 | - | 6, 7 | 700 | |||
| BC048149.1 | - | 3, 4 | 386 | 5706344 | ||
| BC049155.1 | - | 2, 3 | 217 | |||
| CB575943.1 | - | 3, 4 | 281 | |||

Fbxo34
Genomic Map