siRNA Details
| Gene Symbol: Lrrc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142728.1 | NP_001136200.1 | 3 | 555 | ||
| NM_001142729.1 | NP_001136201.1 | 3 | 474 | |||
| NM_030070.3 | NP_084346.2 | 3 | 555 | |||
| GenBank mRNA | AK015292.1 | - | 3 | 473 | ||
| AK019545.1 | - | 3 | 488 | |||
| AK029802.1 | - | 3 | 556 | |||
| AK031279.1 | - | 3 | 474 | |||
| AK039037.1 | - | 3 | 365 | |||
| AK082549.1 | - | 3 | 482 | |||

Lrrc9
Genomic Map