siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 4, 5 | 413 | ||
| GenBank mRNA | AK021042.1 | - | 4, 5 | 304 | ||
| AK210077.1 | - | 1, 2 | 66 | |||
| AY102701.1 | - | 4, 5 | 335 | |||
| BC027201.1 | - | 4, 5 | 349 | |||
| BC043701.1 | - | 4, 5 | 326 | |||
| BC056435.1 | - | 4, 5 | 322 | 5707801 | ||

Naa35
Genomic Map